Learn More
Ace Glass, Inc. Tube, 125mL, with glass stopcock
SDP
Catalog No. 50974796
Encompass_Preferred
125mL standard collecting tube with glass stopcock plug and capillary tubing for connecting to rubber tubing and syringe sampling port with septum.
Catalog No. 50-974-796 Supplier Ace Glass, Inc. Supplier No. 740118
Add to Cart
Add to Cart
125mL standard collecting tube with glass stopcock plug and capillary tubing for connecting to rubber tubing and syringe sampling port with septum.