Learn More
MTC Bio Test Tube / Culture Tube, 12x60mm, 3mL, PP, rimless, no cap, non-sterile, 10 bags of 500 tubes, 5000/cs
Catalog No. 502268493
Encompass_Preferred
MTC Bio’s test tubes are packaged for high volume users in bulk quantity economy cases. Supplied non-sterile and without caps (available separately), all three sizes are available in translucent polypropylene or crystal clear polystyrene. Plug-type caps are available separately. RNase, DNase, pyrogen, & endotoxin free.
This item is not returnable. View return policy
Catalog No. 50-226-8493 Supplier MTC Bio Supplier No. T1121
MTC Bio’s test tubes are packaged for high volume users in bulk quantity economy cases. Supplied non-sterile and without caps (available separately), all three sizes are available in translucent polypropylene or crystal clear polystyrene. Plug-type caps are available separately. RNase, DNase, pyrogen, & endotoxin free.