Learn More
Labcon North America SuperClear 12x75mm Culture Tubes, Polypropylene, in Bulk
Catalog No. 50101511
Encompass_Preferred
Tube, Culture; Labcon; SuperClear; 5mL; 12x75mm culture tubes; For cell culture and general laboratory use; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 3,000 x G; Freezable -80C; Autoclavable; Capacity: 5mL; Seal with plug or dual position caps sold separately; 3000 tubes per bag/1 bag per case; 12mm W. 75mm H; Made in USA
Catalog No. 50-101-511 Supplier Labcon North America Supplier No. 33068000009
Tube, Culture; Labcon; SuperClear; 5mL; 12x75mm culture tubes; For cell culture and general laboratory use; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 3,000 x G; Freezable -80C; Autoclavable; Capacity: 5mL; Seal with plug or dual position caps sold separately; 3000 tubes per bag/1 bag per case; 12mm W. 75mm H; Made in USA