Learn More
Labcon North America SuperClear 12x75mm Culture Tubes with Plug Caps, Polystyrene, 25 per Bag, Sterile
Catalog No. 50101522
Encompass_Preferred
Tube, Culture; Labcon; SuperClear; 5mL; Sterile; 12x75mm culture tubes with attached plug style polyethylene caps; For cell culture and general laboratory use; With extra thick walls to prevent breakage; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polystyrene USP Classs VI resin; Centrifuging to 1,500 x G; Not Freezable; Not Autoclavable; Capacity: 5mL; 25 tubes per bag/20 bags per case; 12mm W. 75mm H; Made in USA
Catalog No. 50-101-522 Supplier Labcon North America Supplier No. 33423350009
Tube, Culture; Labcon; SuperClear; 5mL; Sterile; 12x75mm culture tubes with attached plug style polyethylene caps; For cell culture and general laboratory use; With extra thick walls to prevent breakage; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polystyrene USP Classs VI resin; Centrifuging to 1,500 x G; Not Freezable; Not Autoclavable; Capacity: 5mL; 25 tubes per bag/20 bags per case; 12mm W. 75mm H; Made in USA