Learn More
MTC Bio Tube, 12 x 75mm PP w/cap, 25/bag, sterile
Catalog No. 501127263
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
These disposable sterile tubes are made of high quality polypropylene (PP) plastic that will withstand temperatures from -190 to +120C. They are also available in crystal clear polystyrene (PS) with a range of 0 to 70C. Caps are made of polyethylene (PE) and have the popular two position design (up/loose for aerobic work and down/sealed for anaerobic cultures). Polystyrene tubes may be centrifuged at up to 2000xg. Polypropylene tubes withstand higher forces, up to 4000xg.
Catalog No. 50-112-7263 Supplier MTC Bio Supplier No. T8301
This item has a minimum qty of 2 per supplier requirements.
These disposable sterile tubes are made of high quality polypropylene (PP) plastic that will withstand temperatures from -190 to +120C. They are also available in crystal clear polystyrene (PS) with a range of 0 to 70C. Caps are made of polyethylene (PE) and have the popular two position design (up/loose for aerobic work and down/sealed for anaerobic cultures). Polystyrene tubes may be centrifuged at up to 2000xg. Polypropylene tubes withstand higher forces, up to 4000xg.