Learn More
MTC Bio Transport/ Culture Tube, 16 x 100mm, PP, self-standing, with screw-cap, white printed graduations, 14mL, sterile, 10 bags of 100 tubes, 1000/cs
Catalog No. 502268500
Encompass_Preferred
These 16 x 100 mm screw cap test tubes are available in transparent polypropylene with a skirted self-standing bottom or in crystal clear polystyrene with a traditional round bottom. Each tube has easy to read graduations and a frosted white marking area (12x50mm). Packed 100 tubes (with attached caps) per sterile bag, 10 bags per unit. RNase, DNase, pyrogen, & endotoxin free.
This item is not returnable. View return policy
Catalog No. 50-226-8500 Supplier MTC Bio Supplier No. T7950
These 16 x 100 mm screw cap test tubes are available in transparent polypropylene with a skirted self-standing bottom or in crystal clear polystyrene with a traditional round bottom. Each tube has easy to read graduations and a frosted white marking area (12x50mm). Packed 100 tubes (with attached caps) per sterile bag, 10 bags per unit. RNase, DNase, pyrogen, & endotoxin free.