Learn More
Ace Glass, Inc. Tube, pressure, 15mL, 150psi, 13mm OD, 30.5cm long, complete with#7 front-seal plug
SDP
Supplier:  Ace Glass, Inc. 864853
Encompass_Preferred
15mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Includes #7 front-seal plug. Dimensions: 13mm O.D. x 30.5cm long.
Catalog No. 50-976-174