Learn More
Vee Gee Heathrow Scientific Tube 15ml Bulk Sterile CS500
Catalog No. 502886380
Encompass_Preferred
These conical tubes feature a writing area and flat polyethylene lids for easy labeling.* Lot-to-lot traceability with easy tear packaging* Writing area and flat polyethylene lids for easy labelling* Imprinted graduations and a self-standing version (only in clear bulk pack)* Recyclable chipboard used for the racked versions, making them more environmentally friendly* Skirted
Catalog No. 50-288-6380 Supplier Vee Gee Supplier No. HS4426B
These conical tubes feature a writing area and flat polyethylene lids for easy labeling.* Lot-to-lot traceability with easy tear packaging* Writing area and flat polyethylene lids for easy labelling* Imprinted graduations and a self-standing version (only in clear bulk pack)* Recyclable chipboard used for the racked versions, making them more environmentally friendly* Skirted