Learn More
Thomas Scientific 15mL Centrifuge Tube - Bag, Sterile
SDP
Catalog No. 501459074
Encompass_Preferred
Graduations on clear tubes are printed in bold easy-to-read black ink and are accurate to ± 2% 50mL clear tubes include a black printed 50mL graduation line and 1, 2 and 3mL graduations molded into the conical bottom Large white marking area provides excellent contrast for identification All tubes are manufactured from high quality polypropylene and have smooth, flat surface polyethylene (HDPE) caps Maximum RCF is 12,000g for 15mL conical bottom tubes, 16,000g for 50mL conical bottom tubes, and 6,000g for self-standing tubes Polystyrene tubes Max Force (RCF) is 3,000g Temperature range of -80°C to 120°C 15 and 50mL clear tubes are available in recyclable racks or bulk packaged in zip-closure bags Clear centrifuge tubes are available in sterile (gamma irradiated) or non-sterile pack options NEW! Sure Cap with compressible liner creates a stable seal
Catalog No. 50-145-9074 Supplier Thomas Scientific Supplier No. 1228K62
Graduations on clear tubes are printed in bold easy-to-read black ink and are accurate to ± 2% 50mL clear tubes include a black printed 50mL graduation line and 1, 2 and 3mL graduations molded into the conical bottom Large white marking area provides excellent contrast for identification All tubes are manufactured from high quality polypropylene and have smooth, flat surface polyethylene (HDPE) caps Maximum RCF is 12,000g for 15mL conical bottom tubes, 16,000g for 50mL conical bottom tubes, and 6,000g for self-standing tubes Polystyrene tubes Max Force (RCF) is 3,000g Temperature range of -80°C to 120°C 15 and 50mL clear tubes are available in recyclable racks or bulk packaged in zip-closure bags Clear centrifuge tubes are available in sterile (gamma irradiated) or non-sterile pack options NEW! Sure Cap with compressible liner creates a stable seal