Learn More
Thomas Scientific Denville Centrifuge Tubes, 15ml (17 X 120mm), Molded with Graduations, Polypropylene with screw cap, (Sterile) 25/Bag 500/Qty
SDP
Catalog No. 501458829
Encompass
Printed Graduations (with marking area) New clearer polypropylene copolymer High centrifugal strength - up to 15,000 RCF Flat markable cap with leakproof seal Phenol / Chloroform resistant DNase, RNase and pyrogen free
Catalog No. 50-145-8829 Supplier Thomas Scientific Supplier No. 1158R11
Printed Graduations (with marking area) New clearer polypropylene copolymer High centrifugal strength - up to 15,000 RCF Flat markable cap with leakproof seal Phenol / Chloroform resistant DNase, RNase and pyrogen free