Learn More
Labcon North America 15mL SuperClear Centrifuge Tubes, in Bulk
Catalog No. 50101540
Encompass_Preferred
Tube, Conical centrifuge; Labcon; SuperClear; 15mL; With flat screw caps; Caps include integral molded elastomeric seal; Patented anti-smear writing surface; 3 color printed graduations and reverse reading scale; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 17,000 x G; Freezable -80C; Autoclavable; Capacity: 15mL; Teal color caps; 500 tubes per bag/500 caps in separate bag/500 tubes and caps per case; 17mm W. 118mm H; Made in USA
Catalog No. 50-101-540 Supplier Labcon North America Supplier No. 31418700089
Tube, Conical centrifuge; Labcon; SuperClear; 15mL; With flat screw caps; Caps include integral molded elastomeric seal; Patented anti-smear writing surface; 3 color printed graduations and reverse reading scale; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 17,000 x G; Freezable -80C; Autoclavable; Capacity: 15mL; Teal color caps; 500 tubes per bag/500 caps in separate bag/500 tubes and caps per case; 17mm W. 118mm H; Made in USA