Learn More
Labcon North America 15mL SuperClear Centrifuge Tubes with Plug Style Caps, 25 per Rack, Sterile
Catalog No. 50101537
Encompass_Preferred
Tube, Conical centrifuge; Labcon; SuperClear; 15mL; Sterile; With attached plug style caps; Patented anti-smear writing surface; 3 color printed graduations and reverse reading scale; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polypropylene USP Class VI resin; Centrifuging to 17,000 x G; Freezable -80C; Autoclavable tubes; Capacity: 15mL; Teal color caps; 25 tubes per low waste fiber rack/2 racks per pack/ 10 packs per case; 17mm W. 118mm H; Made in USA
Catalog No. 50-101-537 Supplier Labcon North America Supplier No. 31423350189
Tube, Conical centrifuge; Labcon; SuperClear; 15mL; Sterile; With attached plug style caps; Patented anti-smear writing surface; 3 color printed graduations and reverse reading scale; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polypropylene USP Class VI resin; Centrifuging to 17,000 x G; Freezable -80C; Autoclavable tubes; Capacity: 15mL; Teal color caps; 25 tubes per low waste fiber rack/2 racks per pack/ 10 packs per case; 17mm W. 118mm H; Made in USA