Learn More
Labcon North America 15mL PerformR Polystyrene Centrifuge Tubes, in Bulk Bags
Catalog No. 50101548
Encompass_Preferred
Tube, Conical centrifuge; Labcon; PerformR; 15mL; With flat screw caps, molded graduations, and writing surface; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 3,000 x G; Not Freezable; Not Autoclavable; Capacity: 15mL; Teal color caps; 500 tubes per bag/2 bags per case, caps in separate bag; 17mm W. 118mm H; Made in USA
Catalog No. 50-101-548 Supplier Labcon North America Supplier No. 31158700289
Tube, Conical centrifuge; Labcon; PerformR; 15mL; With flat screw caps, molded graduations, and writing surface; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 3,000 x G; Not Freezable; Not Autoclavable; Capacity: 15mL; Teal color caps; 500 tubes per bag/2 bags per case, caps in separate bag; 17mm W. 118mm H; Made in USA