Learn More
Ace Glass, Inc. Tube, pressure, 170mL, 150psi, 50mm OD, 17.8cm long, complete with #36 back-seal plug
SDP
Catalog No. 50976201
Encompass_Preferred
170mL pressure tube with #36 Ace-Thred, rated for 150psig at as much as 120C. Includes #36 back-seal plug. Dimensions: 50mm O.D. x 17.8cm long.
Catalog No. 50-976-201 Supplier Ace Glass, Inc. Supplier No. 8648122
170mL pressure tube with #36 Ace-Thred, rated for 150psig at as much as 120C. Includes #36 back-seal plug. Dimensions: 50mm O.D. x 17.8cm long.