Learn More
Labcon North America SuperClear 17x100mm Culture Tubes with Dual Position Caps, Polypropylene, 125 per Bag, Sterile
Catalog No. 50101555
Encompass_Preferred
Tube, Culture; Labcon; SuperClear; 14mL; Sterile; 17x100mm culture tubes with attached dual position polyethylene caps; For cell culture and general laboratory use; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polypropylene USP Class VI resin; Centrifuging to 3,000 x G; Freezable -80C; Not Autoclavable; Capacity: 14mL; 125 tubes per bag/8 bags per case; 17mm W. 100mm H; Made in USA
Catalog No. 50-101-555 Supplier Labcon North America Supplier No. 35293550009
Tube, Culture; Labcon; SuperClear; 14mL; Sterile; 17x100mm culture tubes with attached dual position polyethylene caps; For cell culture and general laboratory use; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polypropylene USP Class VI resin; Centrifuging to 3,000 x G; Freezable -80C; Not Autoclavable; Capacity: 14mL; 125 tubes per bag/8 bags per case; 17mm W. 100mm H; Made in USA