Learn More
Labcon North America SuperClear 17x100mm Culture Tubes with Separate Dual Position Caps, Polypropylene, in Bags
Catalog No. 50101557
Encompass_Preferred
Tube, Culture; Labcon; SuperClear; 14mL; 17x100mm culture tubes with separate dual position caps; For cell culture and general laboratory use; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 3,000 x G; Freezable -80C; Not Autoclavable; Capacity: 14mL; Dual position polyethylene caps are in separate bags; 1000 per bag/1 bag per case; 17mm W. 100mm H; Made in USA
Catalog No. 50-101-557 Supplier Labcon North America Supplier No. 35168000009
Tube, Culture; Labcon; SuperClear; 14mL; 17x100mm culture tubes with separate dual position caps; For cell culture and general laboratory use; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 3,000 x G; Freezable -80C; Not Autoclavable; Capacity: 14mL; Dual position polyethylene caps are in separate bags; 1000 per bag/1 bag per case; 17mm W. 100mm H; Made in USA