Learn More
Labcon North America SuperClear 17x100mm Culture Tubes with Separate Dual Position Caps, Polystyrene, in Bags
Catalog No. 50101560
Encompass_Preferred
Tube, Culture; Labcon; SuperClear; 14mL; 17x100mm culture tubes with separate dual position caps; For cell culture and general laboratory use; With extra thick walls to prevent breakage; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polystyrene USP Classs VI resin; Centrifuging to 1,500 x G; Not Freezable; Not Autoclavable; Capacity: 5mL; Dual position polyethylene caps are in separate bags; 1000 per bag/1 bag per case; 12mm W. 75mm H; Made in USA
Catalog No. 50-101-560 Supplier Labcon North America Supplier No. 35158000009
Tube, Culture; Labcon; SuperClear; 14mL; 17x100mm culture tubes with separate dual position caps; For cell culture and general laboratory use; With extra thick walls to prevent breakage; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polystyrene USP Classs VI resin; Centrifuging to 1,500 x G; Not Freezable; Not Autoclavable; Capacity: 5mL; Dual position polyethylene caps are in separate bags; 1000 per bag/1 bag per case; 12mm W. 75mm H; Made in USA