Learn More
Ace Glass, Inc. Tube, pressure, 21mL, 150psi, 19mm OD, 20.3cm long, complete with #7 front-seal plunger valve
SDP
Catalog No. 50976180
Encompass_Preferred
18mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Includes #7 front-seal plunger valve. Dimensions: 19mm O.D. x 20.3cm long.
Catalog No. 50-976-180 Supplier Ace Glass, Inc. Supplier No. 864866
18mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Includes #7 front-seal plunger valve. Dimensions: 19mm O.D. x 20.3cm long.