Learn More
Ace Glass, Inc. Tube, pressure, 2.5mL, 150psi, 8mm OD, 20.3cm long, complete with#7 front-seal plug
SDP
Catalog No. 50974636
Encompass_Preferred
25mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Includes #7 front-seal plug. Dimensions: 8mm O.D. x 20.3cm long.
Catalog No. 50-974-636 Supplier Ace Glass, Inc. Supplier No. 864836
25mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Includes #7 front-seal plug. Dimensions: 8mm O.D. x 20.3cm long.