Learn More
Ace Glass, Inc. Tube, pressure, 200mL, 150psi, 50mm OD, 20.3cm long, complete with #36 front-seal plug
SDP
Catalog No. 50976202
Encompass_Preferred
200mL pressure tube with #36 Ace-Thred, rated for 150psig at as much as 120C. Includes #36 front-seal plug. Dimensions: 50mm O.D. x 20.3cm long.
Catalog No. 50-976-202 Supplier Ace Glass, Inc. Supplier No. 8648123
200mL pressure tube with #36 Ace-Thred, rated for 150psig at as much as 120C. Includes #36 front-seal plug. Dimensions: 50mm O.D. x 20.3cm long.