Learn More
Ace Glass, Inc. Tube, reaction/storage, 200mL, 24/25 standard taper neck, 0-4Hi-Vac valve side arm, No-Air
SDP
Catalog No. 50975355
Encompass_Preferred
200mL Schlenk-style reaction tube for vacuum and No-Air applications. Features a 24/25 standard taper neck, and a 0-4 Hi-Vac valve side arm.
Catalog No. 50-975-355 Supplier Ace Glass, Inc. Supplier No. 7752116
200mL Schlenk-style reaction tube for vacuum and No-Air applications. Features a 24/25 standard taper neck, and a 0-4 Hi-Vac valve side arm.