Learn More
Ace Glass, Inc. Tube, 250mL, with 1:5 PTFE stopcock
SDP
Catalog No. 50979254
Encompass_Preferred
250mL standard collecting tube with 1:5 taper PTFE stopcock plug and capillary tubing for connecting to rubber tubing and syringe sampling port with septum.
Catalog No. 50-979-254 Supplier Ace Glass, Inc. Supplier No. 740150
250mL standard collecting tube with 1:5 taper PTFE stopcock plug and capillary tubing for connecting to rubber tubing and syringe sampling port with septum.