Learn More
Ace Glass, Inc. Tube, pressure, 27mL, 150psi, 19mm OD, 30.5cm long, complete with #7 front-seal plunger valve
SDP
Catalog No. 50976182
Encompass_Preferred
27mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Includes #7 front-seal plunger valve. Dimensions: 19mm O.D. x 30.5cm long.
Catalog No. 50-976-182 Supplier Ace Glass, Inc. Supplier No. 864870
27mL pressure tube with #7 Ace-Thred, rated for 150psig at as much as 120C. Includes #7 front-seal plunger valve. Dimensions: 19mm O.D. x 30.5cm long.