Learn More
Ace Glass, Inc. Tube, pressure, 38mL, 150psi, 25.4mm OD, 20.3cm long, complete with #15 front-seal plunger valve
SDP
Catalog No. 50979801
Encompass_Preferred
38mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 front-seal plunger valve. Dimensions: 25.4mm O.D. x 20.3cm long.
Catalog No. 50-979-801 Supplier Ace Glass, Inc. Supplier No. 864882
38mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 front-seal plunger valve. Dimensions: 25.4mm O.D. x 20.3cm long.