Learn More
Ace Glass, Inc. Tube, reaction/storage, 40mL, 14/20 standard taper neck, 0-3inlet valve side arm, No-Air
SDP
Catalog No. 50979440
Encompass_Preferred
40mL Schlenk-style reaction tube for vacuum and No-Air applications. Features a 14/20 standard taper neck, and a 0-3 inlet valve side arm.
Catalog No. 50-979-440 Supplier Ace Glass, Inc. Supplier No. 7752212
40mL Schlenk-style reaction tube for vacuum and No-Air applications. Features a 14/20 standard taper neck, and a 0-3 inlet valve side arm.