Learn More
Ace Glass, Inc. Tube, pressure, 60mL, 150psi, 38.1mm OD, 10.2cm long, complete with #15 front-seal plunger valve
SDP
Catalog No. NC0207507
Encompass_Preferred
60mL glass pressure tube with #15 Ace-Thred, rated at 150psig at as much as 120C. Includes #15 front-seal plunger valve. Dimensions: 38.1mm O.D. x 10.2cm long.
Catalog No. NC0207507 Supplier Ace Glass, Inc. Supplier No. 864876
60mL glass pressure tube with #15 Ace-Thred, rated at 150psig at as much as 120C. Includes #15 front-seal plunger valve. Dimensions: 38.1mm O.D. x 10.2cm long.