Learn More
Thomas Scientific 50mL Bio-Reaction Tube - Bag, Sterile
SDP
Catalog No. 501461499
Encompass_Preferred
Bio-Reaction tubes offer a convenient and economical option for small scale, high density suspension cell culture. Bio-reaction tubes consist of centrifuge tubes with a 0.22μm hydrophobic membrane in the vent cap. These tubes are perfect for biological reaction experiments that require aseptic conditions with free gas exchange. Save money with the ability to culture and centrifuge in one tube Available in 15 and 50mL conical bottom and 50mL self-standing sizes Easy-to-read black graduations and a large white marking area Manufactured from high quality polypropylene Caps have an ultrasonically welded 0.22μm hydrophobic membrane to allow gas exchange Maximum RCF is 12,000g for 15mL tubes, 16,000 for 50mL conical bottom tubes, and 6,000g for self-standing tubes Packaged sterile (gamma irradiated) in zip-closure bags or foam racks NEW - Vent Control Labels. Labels configured to cover 1-5 of the cap vent openings
Catalog No. 50-146-1499 Supplier Thomas Scientific Supplier No. 1219Z37
Bio-Reaction tubes offer a convenient and economical option for small scale, high density suspension cell culture. Bio-reaction tubes consist of centrifuge tubes with a 0.22μm hydrophobic membrane in the vent cap. These tubes are perfect for biological reaction experiments that require aseptic conditions with free gas exchange. Save money with the ability to culture and centrifuge in one tube Available in 15 and 50mL conical bottom and 50mL self-standing sizes Easy-to-read black graduations and a large white marking area Manufactured from high quality polypropylene Caps have an ultrasonically welded 0.22μm hydrophobic membrane to allow gas exchange Maximum RCF is 12,000g for 15mL tubes, 16,000 for 50mL conical bottom tubes, and 6,000g for self-standing tubes Packaged sterile (gamma irradiated) in zip-closure bags or foam racks NEW - Vent Control Labels. Labels configured to cover 1-5 of the cap vent openings