Learn More
Stellar Scientific TUBE,50ML,POLY,SKIRTED,500/CS
SDP
Catalog No. NC2125635
Encompass
50mL Conical Centrifuge Tubes With Self-Standing Base, RNase and DNase Free, STERILE, Bagged, 500/CS
This item is not returnable. View return policy
Catalog No. NC2125635 Supplier Stellar Scientific Supplier No. T50300
50mL Conical Centrifuge Tubes With Self-Standing Base, RNase and DNase Free, STERILE, Bagged, 500/CS