Learn More
Sarstedt Inc TUBE 50ML PP BROWN 300/PK
Supplier:  Sarstedt Inc 62.547.354
Encompass_Preferred
50ml Screen Printed Screw Cap Tube 28x114mm Amber Conical Bottom Graduated with Writing Block Sterile
Catalog No. NC0867608