Learn More
Labcon North America 50mL SuperClear Centrifuge Tubes with Plug Style Caps, 50 per Bag
Catalog No. 50101567
Encompass_Preferred
Tube, Conical centrifuge; Labcon; SuperClear; 50mL; With attached plug style screw caps; Patented anti-smear writing surface; 3 color printed graduations and reverse reading scale; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 20,000 x G; Freezable -80C; Autoclavable; Capacity: 50mL; Teal color caps; 50 tubes per bag/10 bags per case; 29mm W. 115mm H; Made in USA
Catalog No. 50-101-567 Supplier Labcon North America Supplier No. 31823400089
Tube, Conical centrifuge; Labcon; SuperClear; 50mL; With attached plug style screw caps; Patented anti-smear writing surface; 3 color printed graduations and reverse reading scale; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 20,000 x G; Freezable -80C; Autoclavable; Capacity: 50mL; Teal color caps; 50 tubes per bag/10 bags per case; 29mm W. 115mm H; Made in USA