Learn More
Labcon North America 50mL PerformR Freestanding Centrifuge Tubes, in Bags
Catalog No. 50101623
Encompass_Preferred
Tube, Freestanding conical centrifuge; Labcon; PerformR; 50mL; With attached flat screw caps, black side graduations, and white writing surface; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 12,500 x G; Freezable to -80C; Tubes are autoclavable; Capacity: 50mL; Teal color caps; 50 per bag/10 bags per case; Made in USA
Catalog No. 50-101-623 Supplier Labcon North America Supplier No. 30953400089
Tube, Freestanding conical centrifuge; Labcon; PerformR; 50mL; With attached flat screw caps, black side graduations, and white writing surface; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 12,500 x G; Freezable to -80C; Tubes are autoclavable; Capacity: 50mL; Teal color caps; 50 per bag/10 bags per case; Made in USA