Learn More
CELLTREAT Scientific Products TUBE ONLY, 0.5mL Screw Top Micro Tube, Self-Standing, Grip Band, Non-sterile, 1000/CS
SDP
Catalog No. 501535134
Encompass_Preferred
High temperature resistivity range–shatter/crack resistant and ideal for boiling. Easy 3/4 turn to open and close. Caps are available separately in various colors for easy identification and color-coding. All caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match. Grip band tubes feature ribs that provide exceptional gripping surface and lessen movement during centrifugation. Packaged sterile in re-sealable bags.
Catalog No. 50-153-5134 Supplier CELLTREAT Scientific Products Supplier No. 230811
High temperature resistivity range–shatter/crack resistant and ideal for boiling. Easy 3/4 turn to open and close. Caps are available separately in various colors for easy identification and color-coding. All caps are compatible with all CELLTREAT Screw Top Micro Tubes; mix and match. Grip band tubes feature ribs that provide exceptional gripping surface and lessen movement during centrifugation. Packaged sterile in re-sealable bags.