Learn More
Thomas Scientific Screw Cap Tube, 5mL, Sterile, Racked, Opaque Black
SDP
Catalog No. 501457730
Encompass_Preferred
MTC Bio’s 5mL Five-O™ MacroTubes™ take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption. Useful from -86°C to 110°C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from gamma sterilized racked packaging with attached caps or non-sterile bulk packaging with separate caps. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space. Compatible with standard equipment Centrifuge at up to 25,000xg RNase, DNase & pyrogen free Unsurpassed Clarity Dual graduations at every 0.2mL and 0.5mL
Catalog No. 50-145-7730 Supplier Thomas Scientific Supplier No. 1158Q88
MTC Bio’s 5mL Five-O™ MacroTubes™ take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption. Useful from -86°C to 110°C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from gamma sterilized racked packaging with attached caps or non-sterile bulk packaging with separate caps. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space. Compatible with standard equipment Centrifuge at up to 25,000xg RNase, DNase & pyrogen free Unsurpassed Clarity Dual graduations at every 0.2mL and 0.5mL