Learn More
Ace Glass, Inc. Tube, pressure, 60mL, 150psi, 38.1mm OD, 10.2cm long, complete with #15 back-seal plug
SDP
Catalog No. 50976188
Encompass_Preferred
60mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 back-seal plug. Dimensions: 38.1mm O.D. x 10.2cm long.
Catalog No. 50-976-188 Supplier Ace Glass, Inc. Supplier No. 864898
60mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 back-seal plug. Dimensions: 38.1mm O.D. x 10.2cm long.