Learn More
Ace Glass, Inc. Tube, pressure, 60mL, 150psi, 25.4mm OD, 30.5cm long, complete with #15 front-seal plunger valve
SDP
Supplier:  Ace Glass, Inc. 864885
Encompass_Preferred
60mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120C. Includes #15 front-seal plunger valve. Dimensions: 25.4mm O.D. x 30.5cm long.
Catalog No. 50-976-186