Learn More
Sarstedt Inc Multi-purpose container, 70 ml, (ØxH): 44 x 55 mm, graduated, PP, 500/pack
Catalog No. NC1492531 Shop All Sarstedt Inc. Products
Encompass_Preferred
Tube 70 ml is made of transparent polypropylene and comes with assembled yellow cap.
Catalog No. NC1492531 Supplier Sarstedt Inc Supplier No. 75.9922.744
Add to Cart
Add to Cart
Tube 70 ml is made of transparent polypropylene and comes with assembled yellow cap.