Learn More
Thomas Scientific ThermoGridT .2mL Thermal Cycing Tubes, Strips of 8 tubes and separate strips of 8 caps, red, (Q-PCR) 120 strips per pkg.
SDP
Catalog No. 501458924
Encompass_Preferred
Fits Most Popular Thermal Cyclers RNase/DNase and Pyrogen Free Caps Included Strips of 8 or strips of 12 96 well format Available in Red, Green, Blue, or Yellow Autoclave Safe
Catalog No. 50-145-8924 Supplier Thomas Scientific Supplier No. 1158T79
Fits Most Popular Thermal Cyclers RNase/DNase and Pyrogen Free Caps Included Strips of 8 or strips of 12 96 well format Available in Red, Green, Blue, or Yellow Autoclave Safe