Learn More
Ace Glass, Inc. Tube, pressure, 8mL, 150psi, 13mm OD, 17.8cm long, complete with#7 front-seal plug
SDP
Catalog No. 50974633
Encompass_Preferred
8mL glass pressure tube with #7 Ace-Thred, rated at 150psig at as much as 120C. Includes #7 front-seal plug. Dimensions: 13mm O.D. x 17.8cm long.
Catalog No. 50-974-633 Supplier Ace Glass, Inc. Supplier No. 864825
8mL glass pressure tube with #7 Ace-Thred, rated at 150psig at as much as 120C. Includes #7 front-seal plug. Dimensions: 13mm O.D. x 17.8cm long.