Learn More
Ace Glass, Inc. Tube, pressure, 90mL, 150psi, 50mm OD, 10.2cm long, complete with#36 back-seal plug
SDP
Catalog No. 50979806
Encompass_Preferred
90mL pressure tube with #36 Ace-Thred, rated for 150psig at as much as 120C. Includes #36 back-seal plug. Dimensions: 50mm O.D. x 10.2cm long.
Catalog No. 50-979-806 Supplier Ace Glass, Inc. Supplier No. 8648118
90mL pressure tube with #36 Ace-Thred, rated for 150psig at as much as 120C. Includes #36 back-seal plug. Dimensions: 50mm O.D. x 10.2cm long.