Learn More
Waters Corp Waters Tubing Assembly, Accumulator A Transducer to Vent Valve P3, 10,000 psi, nanoACQUITY System
Catalog No. 50791924 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for the nanoACQUITY system, providing a connection from the accumulator A transducer to port 3 of the vent valve. It is engineered to withstand pressures up to 10,000 psi.
  • Connects the accumulator A transducer to port 3 of the vent valve.
  • Designed for use with 10,000 psi nanoACQUITY systems.
  • Provided as a single unit per package.
  • Recommended replacement as required.
Catalog No. 50-791-924 Supplier Waters Corp Supplier No. 430001534
This tubing assembly is designed for the nanoACQUITY system, providing a connection from the accumulator A transducer to port 3 of the vent valve. It is engineered to withstand pressures up to 10,000 psi.
  • Connects the accumulator A transducer to port 3 of the vent valve.
  • Designed for use with 10,000 psi nanoACQUITY systems.
  • Provided as a single unit per package.
  • Recommended replacement as required.