Learn More
Waters Corp Waters Tubing Assembly, Transducer to Vent Valve P6, 10,000psi nanoACQUITY
Catalog No. 50791925 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly connects the Accumulator B transducer to port 6 of the vent valve, designed for seamless integration into 10,000psi nanoACQUITY systems.
  • Connects the Accumulator B tube from the transducer to port 6 of the vent valve.
  • Designed for use with the 10,000psi nanoACQUITY system.
  • Product type: Spare parts kits.
  • Units per package: 1 per package.
  • Replacement schedule: As required.
Catalog No. 50-791-925 Supplier Waters Corp Supplier No. 430001535
This tubing assembly connects the Accumulator B transducer to port 6 of the vent valve, designed for seamless integration into 10,000psi nanoACQUITY systems.
  • Connects the Accumulator B tube from the transducer to port 6 of the vent valve.
  • Designed for use with the 10,000psi nanoACQUITY system.
  • Product type: Spare parts kits.
  • Units per package: 1 per package.
  • Replacement schedule: As required.