Learn More
Cole-Parmer Cole-Parmer VapLock™ Fitting, Blue PP w/ Yellow ETFE, Straight, Compression to Threaded Adapter, 1/8" OD x 1/4-28 UNF(M); 10/PK
Catalog No. NC1413937 Shop All Cole-Parmer Products
Encompass
Adapters for semi-rigid tubing are color coded by tubing size for easy identification. A two-piece nut and ferrule system provides a liquid-tight connection and withstands pressures up to 500 psi.
Catalog No. NC1413937 Supplier Cole-Parmer Supplier No. 1202008
Adapters for semi-rigid tubing are color coded by tubing size for easy identification. A two-piece nut and ferrule system provides a liquid-tight connection and withstands pressures up to 500 psi.