Learn More
Leica Biosystems Tube attachment with two camera documentation ports for Leica LMD6 (11505223)
Catalog No. NC3366043
Encompass_Preferred
Tube attachment with two camera documentation ports designed for Leica laser microdissection systems. It provides selectable 100:100 manual port distribution to route imaging to dual documentation cameras and is specified for use with LMD6 instruments.
  • Provides two camera documentation ports.
  • Allows manual selection of a 100:100 port distribution.
  • Attaches to the observation tube for imaging.
  • Enables dual-camera documentation workflows for imaging and capture.
Catalog No. NC3366043 Supplier Leica Biosystems Supplier No. 11505223
Tube attachment with two camera documentation ports designed for Leica laser microdissection systems. It provides selectable 100:100 manual port distribution to route imaging to dual documentation cameras and is specified for use with LMD6 instruments.
  • Provides two camera documentation ports.
  • Allows manual selection of a 100:100 port distribution.
  • Attaches to the observation tube for imaging.
  • Enables dual-camera documentation workflows for imaging and capture.