Learn More
Medicus Health Tube Agglutination Viewer, 3"L x 1"W x 0.375"H
SDP
Catalog No. NC1710935
Encompass
Review microagglutination easily with this tube holder ∙Provides a consistent tube height so you don't have to re-adjust the microscope focus ∙Allows you to spin the tube in place to view all areas of the reaction ∙Provides unobstructed views of the cells ∙Holds any size tube in the U-channel ∙3"L x 1"W x 0.375"H
This item is not returnable. View return policy
Catalog No. NC1710935 Supplier Medicus Health Supplier No. 4070M1
Review microagglutination easily with this tube holder ∙Provides a consistent tube height so you don't have to re-adjust the microscope focus ∙Allows you to spin the tube in place to view all areas of the reaction ∙Provides unobstructed views of the cells ∙Holds any size tube in the U-channel ∙3"L x 1"W x 0.375"H