Learn More
Chemglass Life Sciences 25mL Tube, Storage, 14/20 Outer Joint, Airfree, Schlenk
SDP
Catalog No. AF052301
Encompass_Preferred
25mL Tube, Storage, 14/20 Outer Joint, Airfree, Schlenk. Similar to AF-0522, but cylindrical in shape. The 25 and 50mL sizes have a 0-4mm Chem-Vac™ Chem-Cap valve while the 100 and 350mL sizes have a 0-8mm valve size.
Catalog No. AF052301 Supplier Chemglass Life Sciences Supplier No. AF052301
25mL Tube, Storage, 14/20 Outer Joint, Airfree, Schlenk. Similar to AF-0522, but cylindrical in shape. The 25 and 50mL sizes have a 0-4mm Chem-Vac™ Chem-Cap valve while the 100 and 350mL sizes have a 0-8mm valve size.