Learn More
Waters Corp Waters Tubing Assembly, Analytical Column, Union to Outlet
Catalog No. 50791964 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed to connect an analytical column union to the column outlet. It also facilitates the connection of the analytical column outlet to the column out bulkhead.
  • Connects analytical column union to column outlet.
  • Connects analytical column outlet to column out bulkhead (Left side top).
Catalog No. 50-791-964 Supplier Waters Corp Supplier No. 430002235
This tubing assembly is designed to connect an analytical column union to the column outlet. It also facilitates the connection of the analytical column outlet to the column out bulkhead.
  • Connects analytical column union to column outlet.
  • Connects analytical column outlet to column out bulkhead (Left side top).