Learn More
Waters Corp Tubing Assembly, Anal. Inj. Vlv, Port 3, 2545 Pump
Catalog No. 50791962 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly connects an analytical injector valve port 3 to a column inlet. It is specifically designed for use with the 2545 Quaternary Gradient Pump.
  • Connects analytical injector valve port 3 to column inlet.
  • Compatible with 2545 Quaternary Gradient Pump.
  • Sold and shipped only within the US.
  • Available only to end-users.
  • Subject to a tariff surcharge of 8.00%.
Catalog No. 50-791-962 Supplier Waters Corp Supplier No. 430002231
This tubing assembly connects an analytical injector valve port 3 to a column inlet. It is specifically designed for use with the 2545 Quaternary Gradient Pump.
  • Connects analytical injector valve port 3 to column inlet.
  • Compatible with 2545 Quaternary Gradient Pump.
  • Sold and shipped only within the US.
  • Available only to end-users.
  • Subject to a tariff surcharge of 8.00%.