Learn More
Waters Corp Waters Tubing Assembly, Inject Outlet, UPLC, Stainless Steel, .005 in. ID, 11 in.
Catalog No. 50791888 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for use as an inject outlet in ACQUITY UPLC systems. It is constructed from stainless steel and features UPLC fittings on both ends, with dimensions of .005 in. ID by 11 in. long. This part ensures reliable performance in high-speed applications within the UPLC system.
  • Designed for UPLC systems.
  • Constructed from stainless steel.
  • Features .005 in. ID and 11 in. length.
  • Equipped with fittings on both ends.
  • Functions as an inject outlet tube assembly.
  • Available in 1 unit per package.
Catalog No. 50-791-888 Supplier Waters Corp Supplier No. 430001084
This tubing assembly is designed for use as an inject outlet in ACQUITY UPLC systems. It is constructed from stainless steel and features UPLC fittings on both ends, with dimensions of .005 in. ID by 11 in. long. This part ensures reliable performance in high-speed applications within the UPLC system.
  • Designed for UPLC systems.
  • Constructed from stainless steel.
  • Features .005 in. ID and 11 in. length.
  • Equipped with fittings on both ends.
  • Functions as an inject outlet tube assembly.
  • Available in 1 unit per package.