Learn More
Waters Corp Waters Corporation Tubing Assembly, Inlet to PDA, Low Flow, PEEK, 0.0025 mm ID, 6 in.
Catalog No. 50791936 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Inlet Tube Assembly is designed to fit on ACQUITY UPLC TUV Detectors. It is compatible with detectors having serial numbers through K05UPT699N and from K05UPT700N and later.
  • Designed for UPLC systems
  • Made from PEEK material
  • Features an internal diameter of 0.0025 mm
  • Has a length of 6 in.
  • Compatible with ACQUITY UPLC TUV Detectors with serial numbers through K05UPT699N and with serial numbers from K05UPT700N and later
  • Replacement as required
Catalog No. 50-791-936 Supplier Waters Corp Supplier No. 430001749
The Inlet Tube Assembly is designed to fit on ACQUITY UPLC TUV Detectors. It is compatible with detectors having serial numbers through K05UPT699N and from K05UPT700N and later.
  • Designed for UPLC systems
  • Made from PEEK material
  • Features an internal diameter of 0.0025 mm
  • Has a length of 6 in.
  • Compatible with ACQUITY UPLC TUV Detectors with serial numbers through K05UPT699N and with serial numbers from K05UPT700N and later
  • Replacement as required