Learn More
Waters Corp Waters Tubing Kit, Solvent Inlet, ACQUITY I-Class Binary Solvent Manager
Catalog No. 50201657 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a replacement kit of solvent inlet tube assemblies, specifically designed for use with ACQUITY I-Class Binary Solvent Managers. It ensures optimal performance and reliable operation of the system.
  • Replacement kit of solvent inlet tube assemblies
  • Compatible with ACQUITY I-Class Binary Solvent Managers
  • Also suitable for ACQUITY H-Class Binary Solvent Managers
Catalog No. 50-201-657 Supplier Waters Corp Supplier No. 430002800
This product is a replacement kit of solvent inlet tube assemblies, specifically designed for use with ACQUITY I-Class Binary Solvent Managers. It ensures optimal performance and reliable operation of the system.
  • Replacement kit of solvent inlet tube assemblies
  • Compatible with ACQUITY I-Class Binary Solvent Managers
  • Also suitable for ACQUITY H-Class Binary Solvent Managers